Learn More
DuPont™ Tyvek™ 500 HV Coverall w/ Elastic Wrists and Ankles
GREENER_CHOICE
Catalog No. 17000526 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-000-526 Small
17-000-527 Medium
17-000-528 Large
17-000-529 X-Large
17-000-530 2X-Large
17-000-531 3X-Large
17-000-532 4X-Large
17-000-533 5X-Large
17-000-534 6X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 17-000-526 Supplier DuPont™ Supplier No. TY125SHVSM0025XC
Add to Cart
Add to Cart
500 HV garments are mainly composed of flash spun polyethylene, providing an ideal balance of protection, durability and comfort
  • EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination), EN ISO 20471 (High Visibility Clothing, Class 3)
  • Antistatic treatment (EN 1149-5) - on inside
  • Stitched external seams
  • Very low inward leakage thanks to optimized design
  • Tyvek™ auto-lock zipper and zipper flap for increased protection.
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Collared coverall. Mandarin collar
  • Products made after January, 2023 are manufactured under specifications that do not contain natural rubber latex
TRUSTED_SUSTAINABILITY
Specifications
Product Type High Visibility Coveralls
Size Small
Gender Unisex
Material Tyvek
Color Fluorescent Orange, Orange
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Closure Type Zipper
Seam Type Serged
Certifications/Compliance ANSI/ISEA 107-2020 Type R/Class 3 Section 11
Certifications/Compliance (Europe) EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination), EN ISO 20471 (High Visibility Clothing, Class 3), Antistatic treatment (EN 1149-5) on inside