Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keystone Safety Tyvek™ Bouffant Cap
SDP

Catalog No. 19055170
This item is not returnable. View return policy
Catalog No. 19-055-170 Supplier Keystone Safety Supplier No. 110TVKI21
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Microporous Bouffant caps deliver an ideal balance of protection, durability and comfort.

  • Tyvek™ Material
  • 100% Latex Free
  • 21 in.
  • White

Specifications

Product Type Bouffant
Material Tyvek
Color White
Disposable Single-use
Latex Free Yes
Model 110TVK
Sterility Non-sterile
For Use With (Application) Restaurants, Hospitals, Salons, Spas, Catering Events
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.