Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CleanWear™ Tyvek Flask Cover

Catalog No. p-4258150 Shop All Cleanwear USA Products
Encompass
Change view
Click to view available options
Description:
30.48 cm dia.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
18-892-090 30.48 cm dia.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-892-090 Supplier CleanWear™ Supplier No. 9632850
Only null left
Add to Cart
Add to Cart

Specifications

Description 30.48 cm dia.
Diameter (English) Outer 12 in.
For Use With (Equipment) For flask
Product Type Cover, Flask
Diameter (Metric) Outer 30.48 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.