Learn More
DuPont™ IsoClean™ Frock with Front-Snap Closures
Catalog No. 190881136E Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Medium
X-Large
2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-088-1136E 2X-Large
19-088-1136B Medium
19-088-1136D X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Catalog No. 19-088-1136E Supplier DuPont™ Supplier No. IC271BWH2X00300B
This item is not returnable. View return policy
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
No pockets to trap contamination.
Made of Tyvek™, a spunbonded olefin fabric that is lightweight and flexible, yet provides superior contamination control and particulate filtration, unsurpassed liquid and particulate shielding and two-way contamination protection. Garments are constructed with the smooth side out to reduce particulate entrapment and bound seams reduce lint. Processed in a certified ISO Class 4 (formerly Class 10) cleanroom, antistatic treated and packaged in durable, antistatic bags. ISO Class 5 (formerly Class 100) compatible. Constructed to hold up and keep you comfortable. Generous sizing permits freedom of movement. Plastic snaps won't rust or flake during processing. Available in bulk (no special cleaning), clean (ISO Class 4 [formerly Class 10] cleanroom processed), sterile (gamma irradiated), or clean and sterile. Every lot is 100% traceable.
  • Excellent barrier to dry particulates, nonhazardous liquids, and microorganisms
  • Bound neck with adjustable snaps Five snap front Tunneled elastic wrist Made from DuPont Tyvek™ brand protective material Unique, patented flashspinning process creates an excellent barrier to dry particles, microorganisms and nonhazardous liquids Comfortable, lightweight and durable Full traceability on all sterilized apparel Recyclable
TRUSTED_SUSTAINABILITY
Specifications
Size 2X-Large
Material Tyvek
Color White
Sterility Non-sterile
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.