Learn More
DuPont™ Tyvek™ IsoClean™ Series 108 Cleanroom Coveralls, Attached Hood and Boots
BIOPHARMAGREENER_CHOICE
Catalog No. 17988179 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-988-179 Small
17-988-180 Medium
17-988-181 Large
17-988-182 X-Large
17-988-183 2X-Large
17-988-184 3X-Large
17-988-185 4X-Large
17-988-186 5X-Large
17-988-187 6X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 17-988-179 Supplier DuPont™ Supplier No. IC108SWHSM0025TS
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Cleanroom coveralls with attached hood and boots deliver an ideal balance of protection, durability, and comfort.
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam.
  • Coverall has elastic openings for tighter fit at wrist and ankle.
  • Attached hood with elastic around face opening.
  • Attached thumb loops to minimize bunching of sleeves.
  • Full traceability on all sterilized apparel with Certificates of Sterility.
  • Double bagged.
  • Attached boots with PVC soles that provide enhanced skid-resistance and durability.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleanroom Coveralls
Size Small
Gender Unisex
Material Tyvek
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Thumb Loop
Hood Type Respirator Fit
Closure Type Zipper
Seam Type Serged
Sterility Sterile
Description Individually double bagged
Model IC108SWH
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.