Learn More
DuPont™ Tyvek™ IsoClean™ Coverall with Elastic Wrists, Ankles, Waist, Double Bagged
Shop All DuPont Personal Protection ProductsDescription
Advantages of Tyvek™ Isoclean™ garments:
- Unique, patented flash spinning process creates an inherent barrier to dry particles, microorganisms and non-hazardous liquids
- Comfortable, lightweight and durable
- Clean-processed garments offer lowest level of particle shedding within DuPont portfolio
- Sterile garment options are gamma sterilized to an SAL of 10-6
- Both Serged and bound seam with covered elastic options
- Bound seam garments offer highest particle barrier within DuPont™ IsoClean™ product portfolio
- Full traceability on all sterilized apparel
Certifications
Cat. III Type 5-B and 6-B
Specifications
Specifications
| Product Type | Cleanroom Coveralls |
| Size | 2X-Small |
| Gender | Unisex |
| Material | Tyvek |
| Color | White |
| Disposable | Single-use |
| Ankle Style | Elastic |
| Cuff Style | Elastic |
| Neck Style | Collared |
| Seam Type | Bound |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.