Learn More
DuPont™ Tyvek™ IsoClean™ Coverall with Elastic Wrists, Ankles, Waist, Double Bagged
Catalog No. 18888711 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
2X-Small
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
7X-Large
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-888-711 X-Small
18-888-700 2X-Small
18-888-701 2X-Large
18-888-702 3X-Large
18-888-703 4X-Large
18-888-704 5X-Large
18-888-705 6X-Large
18-888-706 7X-Large
18-888-707 Large
18-888-708 Medium
18-888-709 Small
18-888-710 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Catalog No. 18-888-711 Supplier DuPont™ Supplier No. IC183BWHXS0025DS
This item is not returnable. View return policy
DuPont™ Tyvek™ IsoClean™ Coverall with Elastic Wrists delivers an ideal balance of protection, durability and comfort
Advantages of Tyvek™ Isoclean™ garments:
  • Unique, patented flash spinning process creates an inherent barrier to dry particles, microorganisms and non-hazardous liquids
  • Comfortable, lightweight and durable
  • Clean-processed garments offer lowest level of particle shedding within DuPont portfolio
  • Sterile garment options are gamma sterilized to an SAL of 10-6
  • Both Serged and bound seam with covered elastic options
  • Bound seam garments offer highest particle barrier within DuPont™ IsoClean™ product portfolio
  • Full traceability on all sterilized apparel
Certifications
Cat. III Type 5-B and 6-B
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleanroom Coveralls
Size X-Small
Gender Unisex
Material Tyvek
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Seam Type Bound
Sterility Sterile
Resistance Particle
Certifications/Compliance (Europe) Cat. III, Type 5-B, 6-B
Latex Free Yes
Model IC183BWH
Quantity 25/Cs.