Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tyvek™ IsoClean™ 255 Series Coveralls
BIOPHARMAGREENER_CHOICE

Catalog No. 18888110 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-888-110 Small
18-888-111 Medium
18-888-112 Large
18-888-113 X-Large
18-888-114 2X-Large
18-888-115 3X-Large
18-888-116 4X-Large
18-888-117 5X-Large
18-888-118 6X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 18-888-110 Supplier DuPont™ Supplier No. IC255BWHSM0025CS
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sterile coverall with collar for use in ISO 6 (Class 1,000) or higher controlled environments.

  • Tyvek flashspun polyolefin protective material
  • Patented flash-spinning process
  • Creates barrier to dry particles, microorganisms, non-hazardous liquids
  • Lightweight, durable
  • Collared
  • Elastic thumb loops keep sleeves in place
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Coveralls
Size Small
Gender Unisex
Material Tyvek
Color White
Disposable Single-use
Ankle Style Covered Elastic
Hood Type Standard
Closure Type Zipper
Seam Type Bound
Sterility Sterile
Certifications/Compliance (Europe) CE
Model IC255BWH
Sleeve Style Set sleeves
For Use With (Application) Protective Barrier to Dry Particle, Microorganisms
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.