Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tyvek™ IsoClean™ Front Snap Cleanroom Frocks, High Mandarin Collar
GREENER_CHOICE

Catalog No. 17988079 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-988-079 4X-Large
17-988-073 Small
17-988-074 Medium
17-988-075 Large
17-988-076 X-Large
17-988-077 2X-Large
17-988-078 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Catalog No. 17-988-079 Supplier DuPont™ Supplier No. IC263SWH4X00300B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.

Cleanroom frocks feature a high mandarin collar with snap for a secure fit and provides an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash.

  • Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam
  • Elastic opening for tighter fit at wrist
  • Mandarin collar with snap for secure fit
  • Frocks provide body-length jacket design with front snap closures for easy donning and doffing
  • Bulk packaged in double transparent poly liners.
  • Manufactured under specifications that do not contain natural rubber latex
TRUSTED_SUSTAINABILITY

Specifications

Size 4X-Large
Material Tyvek
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Snaps
Cuff Style Elastic
Collar Type High Mandarin
Size (English) Chest 46 to 52 in.
Sleeve Style Set-in Sleeve
Length (Metric) 113.03 cm
Length (English) Sleeve 43.75 in.
Length (Metric) Sleeve 111.12 cm
Cleanroom Class Class 1000 (ISO 6)
Length (English) 44.5 in.
Product Type Controlled Environments Frock
Seam Type Serged
For Use With (Application) Cleanroom, Pharmaceutical, Biotechnology, Medical Device Manufacturing, and Laboratory Environments
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.