Learn More
DuPont Personal Protection Tyvek™ IsoClean™ Sterile Sleeve Cover
Shop All DuPont Personal Protection ProductsDescription
Tyvek™ IsoClean™ delivers an ideal balance of protection, durability and comfort. Made using a patented flash spinning process, Tyvek™ provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash.
- Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
- Elastic at both ends (bicep and wrist) for arm protection
- Incorporated thumb loop for secure positioning on the arm
- 21 in. length
- One size fits most
- Pair packed and folded for ease of aseptic donning
- Case contains double transparent poly liners
- Manufactured under specifications that do not contain natural rubber latex
Specifications
Specifications
| Size | Universal |
| Material | Tyvek |
| Color | White |
| Sterility | Sterile |
| Disposable | Single-use |
| Closure Type | Elastic Ends |
| Seam Type | Bound |
| Latex Free | Yes |
| Includes | Thumb loops |
| Length (English) | 21 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.