Learn More
MSC TYVEK LAB COAT
Catalog No. 5000786832
Encompass
These disposable jackets and coats are designed for protection in environments requiring chemical resistance. They are supplied in a large size and feature a convenient snaps closure, ensuring ease of use and secure wear.
  • Large size
  • Snaps closure
  • Chemical resistant
  • Meets multiple industry standards (e.g., ASTM D774, OSHA 16 CFR 1610)
  • Sleeve length 35-1/4 in
Catalog No. 50-007-86832 Supplier MSC Supplier No. 36782571
These disposable jackets and coats are designed for protection in environments requiring chemical resistance. They are supplied in a large size and feature a convenient snaps closure, ensuring ease of use and secure wear.
  • Large size
  • Snaps closure
  • Chemical resistant
  • Meets multiple industry standards (e.g., ASTM D774, OSHA 16 CFR 1610)
  • Sleeve length 35-1/4 in