Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tyvek ™ Micro-Clean ™ 2-1-2 Coveralls

Catalog No. 17988014 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-988-014 2X-Large
17-988-015 3X-Large
17-988-012 Large
17-988-011 Medium
17-988-013 X-Large
17-989-608 Small
17-989-609 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 17-988-014 Supplier DuPont™ Supplier No. CC252BBU2X0025PI
This item is not returnable. View return policy

Ideal for controlled environments to protect against micro-organisms and particulates.

Coveralls feature Micro-Clean™ with 2-1-2 polymeric resin coating that allows for thorough protection and reduction in static.

  • Made from Tyvek™ flash spun polyolefin protective material to create a barrier to particulates, nonhazardous liquids and micro-organisms
  • Available clean processed (individually packaged)
  • Coated in 2-1-2 polymeric resin
  • Antistatic
  • Bound neck and seams with zipper closure
  • Dolman sleeves
  • Elastic wrists and ankles

TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Coveralls
Size 2X-Large
Material Tyvek Micro-Clean Flashspun Polyethylene
Color Blue
Disposable Single-use
Ankle Style Covered Elastic
Cuff Style Covered Elastic
Neck Style Collared
Closure Type Zipper
Seam Type Bound
Sterility Non-sterile
Coating Polymeric Resin
Collar Type Bound
Size (English) Chest 43 to 46 in.
Size (English) Inseam 30.75 in.
Size (Metric) Chest 109.2 to 116.84 cm
Size (Metric) Inseam 78 cm
Sleeve Style Dolman
Thickness 5 mil
For Use With (Application) Barrier to dry particles and microorganisms
Length (English) Sleeve 37.75 in.
Length (Metric) Sleeve 95.8 cm
Height (English) 6.1 to 6.5 ft.
Height (Metric) 185.92 to 198.12 cm
Cleanroom Class Class 1000 (ISO 6)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.