Learn More
Description
Coveralls feature Micro-Clean™ with 2-1-2 polymeric resin coating that allows for thorough protection and reduction in static.
- Made from Tyvek™ flash spun polyolefin protective material to create a barrier to particulates, nonhazardous liquids and micro-organisms
- Available clean processed (individually packaged)
- Coated in 2-1-2 polymeric resin
- Antistatic
- Bound neck and seams with zipper closure
- Dolman sleeves
- Elastic wrists and ankles
Specifications
Specifications
| Product Type | Cleanroom Coveralls |
| Size | 3X-Large |
| Material | Tyvek Micro-Clean Flashspun Polyethylene |
| Color | Blue |
| Disposable | Single-use |
| Ankle Style | Covered Elastic |
| Cuff Style | Covered Elastic |
| Neck Style | Collared |
| Closure Type | Zipper |
| Seam Type | Bound |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.