Learn More
HPK INDUSTRIES Muff Cover Fits a 4 Inch Opening, Tyvek Material
Catalog No. NC1610294
Encompass_Preferred
HPK covers provide a convenient means to cover bowls, flanges, flasks, beakers, containers, media bottles, and hoses. Tyvek materials are breathable, so when covering wet parts and equipment, moisture can still evaporate. They are puncture resistant and extremely tough. Each closure incorporates latex-free 1/4″ elastic to create a positive seal around all openings.
Catalog No. NC1610294 Supplier HPK INDUSTRIES Supplier No. 310304B
HPK covers provide a convenient means to cover bowls, flanges, flasks, beakers, containers, media bottles, and hoses. Tyvek materials are breathable, so when covering wet parts and equipment, moisture can still evaporate. They are puncture resistant and extremely tough. Each closure incorporates latex-free 1/4″ elastic to create a positive seal around all openings.