Learn More
MSC Dupont Tyvek Work Pants, 3X-Large, White, Serged Seams, 50 Pack
Catalog No. 5000770234
Encompass
Tyvek 400 garments, made from flash-spun high-density polyethylene, offer a balance of protection, durability, and comfort. They provide an inherent barrier against particles down to 1.0 micron, making them suitable for applications like lead and asbestos abatement, general maintenance, and spray painting.
  • Inherent barrier against particles down to 1.0 micron
  • Protection built into fabric, no films or laminates to wear away
  • Durable for consistent barrier protection after wear and abrasion
  • Open ankle and elastic waist for easy donning and removal
  • Disposable design
  • Features serged seams
Catalog No. 50-007-70234 Supplier MSC Supplier No. 05671474
Tyvek 400 garments, made from flash-spun high-density polyethylene, offer a balance of protection, durability, and comfort. They provide an inherent barrier against particles down to 1.0 micron, making them suitable for applications like lead and asbestos abatement, general maintenance, and spray painting.
  • Inherent barrier against particles down to 1.0 micron
  • Protection built into fabric, no films or laminates to wear away
  • Durable for consistent barrier protection after wear and abrasion
  • Open ankle and elastic waist for easy donning and removal
  • Disposable design
  • Features serged seams