Learn More
Andwin Scientific TYVEX APRONS WHITE 100/CS
SDP
Catalog No. NC0698592
Encompass_Preferred
The apron is made with DuPont Tyvek material that allows this product to combat against penetration and to be chemically and biologically inert. This product has a bib style and is 91.4 centimeters or 36 inches long. It is made to fit anyone as it has a universal size. This product is sold in a case of 100 aprons in the color of white.
Catalog No. NC0698592 Supplier Andwin Scientific Supplier No. 80080916
The apron is made with DuPont Tyvek material that allows this product to combat against penetration and to be chemically and biologically inert. This product has a bib style and is 91.4 centimeters or 36 inches long. It is made to fit anyone as it has a universal size. This product is sold in a case of 100 aprons in the color of white.