Learn More
YSI 3P LifeScience 20 mL Dosing Unit
Catalog No. NC1045903
Encompass
20 mL complete dosing unit for the TITRONIC 300. Includes piston, piston rod and UV protection coat. This item is included with T 300/20 M1 and T 300/20 M2.
Catalog No. NC1045903 Supplier YSI 3P LifeScience Supplier No. 285223718
20 mL complete dosing unit for the TITRONIC 300. Includes piston, piston rod and UV protection coat. This item is included with T 300/20 M1 and T 300/20 M2.