Learn More
Andwin Scientific Brother Black/White PET Label Tape Cartridge 12MM
SDP
Catalog No. NC0471561
Encompass_Preferred
Brother Black/White PET Label Tape Cartridge can be utilized in hot and cold environments, both indoor and outdoor. This can be used for everyday general labeling needs. It is best applied on smooth, flat surfaces.
Catalog No. NC0471561 Supplier Andwin Scientific Supplier No. 13C490
Brother Black/White PET Label Tape Cartridge can be utilized in hot and cold environments, both indoor and outdoor. This can be used for everyday general labeling needs. It is best applied on smooth, flat surfaces.