Learn More
MSC Brother Laminated Label Tape, Black/Red, 12 mm, 1 ea
Catalog No. NC3314785
Encompass
This laminated label tape is designed for use with Brother P-touch label makers, providing durable and legible labels for various indoor and outdoor applications.
  • Black print on red tape for clear visibility.
  • Six-layer laminated construction ensures durability against fading and peeling.
  • Standard adhesive suitable for smooth, flat surfaces.
  • Split back design allows for easy peeling and application.
  • Compatible with P-touch label makers accepting 12 mm TZe label tape.
  • Measures 12 mm wide and 8 m long.
Catalog No. NC3314785 Supplier MSC Supplier No. 73859944
This laminated label tape is designed for use with Brother P-touch label makers, providing durable and legible labels for various indoor and outdoor applications.
  • Black print on red tape for clear visibility.
  • Six-layer laminated construction ensures durability against fading and peeling.
  • Standard adhesive suitable for smooth, flat surfaces.
  • Split back design allows for easy peeling and application.
  • Compatible with P-touch label makers accepting 12 mm TZe label tape.
  • Measures 12 mm wide and 8 m long.