Learn More
TA Instruments Tzero Hermetic Alodined Lids, Aluminum Hermetic Lids, Use with Tzero Alodined Pans, Hermetic Encapsulation, Moisture or Solvent Containing Materials, Reliable Sealing, Preserve Sample Integrity, DSC and TGA Applications, 100 Pack
Catalog No. 5000580908 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Tzero Hermetic Alodined Lids are aluminum hermetic lids for use with Tzero Alodined Pans in hermetic encapsulation of materials containing moisture or solvents. Sample encapsulation requires the Tzero Sample Press Kit. They provide reliable sealing for DSC and TGA applications and preserve sample integrity during thermal analysis.
Catalog No. 50-005-80908 Supplier TA Instruments Supplier No. 901670.901
Waters TA Instruments Tzero Hermetic Alodined Lids are aluminum hermetic lids for use with Tzero Alodined Pans in hermetic encapsulation of materials containing moisture or solvents. Sample encapsulation requires the Tzero Sample Press Kit. They provide reliable sealing for DSC and TGA applications and preserve sample integrity during thermal analysis.