Learn More
TA Instruments Tzero Powder Sample Preparation Kit, Designed for Powdered Samples, Provides Non Skid Base, Stainless Steel Tools and Optimized Loading for Reliable DSC Results
Catalog No. 5000580715 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Tzero Powder Sample Preparation Kit is designed for users working with powdered samples and Tzero pans. It includes a non skid base and three part stainless steel tool set to simplify loading. The design optimizes sample placement, prevents contamination, and ensures reliable, repeatable DSC results with powdered materials.
Catalog No. 50-005-80715 Supplier TA Instruments Supplier No. 972800.901
Waters TA Instruments Tzero Powder Sample Preparation Kit is designed for users working with powdered samples and Tzero pans. It includes a non skid base and three part stainless steel tool set to simplify loading. The design optimizes sample placement, prevents contamination, and ensures reliable, repeatable DSC results with powdered materials.