Learn More
TA Instruments Tzero Sample Press, for Sealing or Crimping DSC and TGA Sample Pans, Pan Specific Die Set Required Separately
Catalog No. 5000580708 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Tzero Sample Press is used for sealing or crimping Waters TA Instruments DSC and TGA sample pans without a pan-specific die set. A separately sold die set is required for sealing and crimping pans.
Catalog No. 50-005-80708 Supplier TA Instruments Supplier No. 956961.901
Waters TA Instruments Tzero Sample Press is used for sealing or crimping Waters TA Instruments DSC and TGA sample pans without a pan-specific die set. A separately sold die set is required for sealing and crimping pans.