Learn More
Enzo Life Sciences U-0126 (1mg). CAS: 109522-58-2
SDP
Catalog No. 502010665
Encompass_Preferred
U-0126 (Bis[amino[(2-aminophenyl)thio]methylene]butanedinitrile) is a novel, potent and selective MEK inhibitor (MEK1; IC50=72nM, MEK2; IC50=58nM). Inhibits AP-1 transcription activity. Blocks the production of inflammatory cytokines and matrix metalloproteinases. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (>10mg/ml) or 100% ethanol (2mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0665 Supplier Enzo Life Sciences Supplier No. BMLEI2820001
U-0126 (Bis[amino[(2-aminophenyl)thio]methylene]butanedinitrile) is a novel, potent and selective MEK inhibitor (MEK1; IC50=72nM, MEK2; IC50=58nM). Inhibits AP-1 transcription activity. Blocks the production of inflammatory cytokines and matrix metalloproteinases. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (>10mg/ml) or 100% ethanol (2mg/ml). Long Term Storage: Ambient.