Learn More
Enzo Life Sciences U-0521 (100mg). CAS: 5466-89-7
SDP
Supplier:  Enzo Life Sciences BMLEI2420100
Encompass_Preferred
Catechol-O-methyltransferase (COMT) inhibitor. Alternative name: 3' 4'-Dihydroxyisobutyrophenone. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (25 mg/ml) or ethanol (25 mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0630