Learn More
Selleck Chemical LLC U-104 10mg 178606-66-1 MST-104, NSC 213841, SLC-0111, WBI-5111
SDP
Supplier:  Selleck Chemical LLC S286610MG
Encompass_Preferred
U-104 (MST-104, NSC 213841, SLC-0111, WBI-5111) is a potent carbonic anhydrase (CA) inhibitor for CA IX and CA XII with Ki of 45.1 nM and 4.5 nM, respectively, very low inhibition for CA I and CA II. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3103