Learn More
Cole-Parmer Idex U-116 Chromatography Tubing, 316 SS, 1/16" OD x 0.030" ID x 4" L, White; 1/EA
Catalog No. NC3234198 Shop All Cole-Parmer Products
Encompass
Precut stainless steel tubing ready for all chromatography testing requirements316 stainless steel tubing, polished and passivatedColor coded for easy identificationSuited for most analytical applicationsMax pressure 12,000 psi1 year warranty
Catalog No. NC3234198 Supplier Cole-Parmer Supplier No. 0201121
Precut stainless steel tubing ready for all chromatography testing requirements316 stainless steel tubing, polished and passivatedColor coded for easy identificationSuited for most analytical applicationsMax pressure 12,000 psi1 year warranty