Learn More
Enzo Life Sciences Pregnenolone-16alpha-carbonitrile (20 mg). CAS: 1434-54-4
SDP
Supplier:  Enzo Life Sciences BMLS6200020
Encompass_Preferred
Cytochrome P-450 inducer. Activates transcription of the PCN1 gene. SXR/PXR Ligand. Alternative name: U-14975. Purity: ≥99% (HPLC). Solubility: Soluble in DMF (5mg/ml) or acetone (1mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-1751