Learn More
Cell Signaling Technology U-18666A, Cholesterol Transport Inhibitor, 5 mg
SDP
Catalog No. 502050551
Encompass
U-18666A is a cell-permeable small molecule that functions as a potent inhibitor of intracellular cholesterol transport. By blocking low-density lipoprotein (LDL)-derived cholesterol trafficking, this compound prevents cholesterol esterification and suppresses LDL receptor activity. It is widely utilized in research to simulate intracellular cholesterol accumulation, providing a valuable tool for investigating the pathogenesis of neurological disorders, such as Alzheimer™ disease and Niemann-Pick disease type C, as well as atherosclerosis and viral infection mechanisms.
  • Inhibits LDL-derived cholesterol transport and esterification
  • Suppresses LDL receptor activities
  • Useful for studying intracellular cholesterol accumulation and neurodegenerative pathologies
  • Relevant for research on viral entry and replication
  • Supplied as a lyophilized powder
Catalog No. 50-205-0551 Supplier Cell Signaling Technology Supplier No. 68998S
U-18666A is a cell-permeable small molecule that functions as a potent inhibitor of intracellular cholesterol transport. By blocking low-density lipoprotein (LDL)-derived cholesterol trafficking, this compound prevents cholesterol esterification and suppresses LDL receptor activity. It is widely utilized in research to simulate intracellular cholesterol accumulation, providing a valuable tool for investigating the pathogenesis of neurological disorders, such as Alzheimer™ disease and Niemann-Pick disease type C, as well as atherosclerosis and viral infection mechanisms.
  • Inhibits LDL-derived cholesterol transport and esterification
  • Suppresses LDL receptor activities
  • Useful for studying intracellular cholesterol accumulation and neurodegenerative pathologies
  • Relevant for research on viral entry and replication
  • Supplied as a lyophilized powder