Learn More
CYTION U-343 MG Cells, 1 cryovial, Organism- Human, Tissue- Brain, Disease- Glioblastoma
Catalog No. NC3245530
Encompass
The U-343 MG cell line is derived from a human glioblastoma, a type of aggressive brain tumor. Originally isolated from a 54-year-old Caucasian male, this cell line has been widely used in neurological research, particularly in studies concerning the pathology and therapeutic treatment strategies for glioblastoma. The U-343 MG cell line is notable for its astrocytic properties, resembling those of astrocytes in the brain, which makes it particularly useful for studying tumor behavior and neurobiology in a controlled in vitro environment.Genetically, the U-343 MG cells are characterized by various mutations typical of glioblastoma, including alterations in the TP53 gene and the EGFR gene. These mutations not only offer insights into the molecular underpinnings of glioblastoma malignancy but also serve as potential targets for therapeutic intervention.
Catalog No. NC3245530 Supplier CYTION Supplier No. 300365
The U-343 MG cell line is derived from a human glioblastoma, a type of aggressive brain tumor. Originally isolated from a 54-year-old Caucasian male, this cell line has been widely used in neurological research, particularly in studies concerning the pathology and therapeutic treatment strategies for glioblastoma. The U-343 MG cell line is notable for its astrocytic properties, resembling those of astrocytes in the brain, which makes it particularly useful for studying tumor behavior and neurobiology in a controlled in vitro environment.Genetically, the U-343 MG cells are characterized by various mutations typical of glioblastoma, including alterations in the TP53 gene and the EGFR gene. These mutations not only offer insights into the molecular underpinnings of glioblastoma malignancy but also serve as potential targets for therapeutic intervention.