Learn More
Enzo Life Sciences U-37883A (5mg). CAS: 57568-80-6
SDP
Supplier:  Enzo Life Sciences BMLKC1590005
Encompass_Preferred
A non-sulfonylurea inhibitor of K-ATP channels. Inhibits vascular K-ATP channels. Inhibits dopamine-modulated potassium channels. Alternative name: N-(1-Adamantyl)-N'-cyclohexyl-4-morpholinecarboxamidine . HCl. Purity: ≥98% (TLC). Solubility: Soluble in water (5mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-1073