Learn More
Abcam U-37883A, ATP-sensitive K+ channel blocker, 5MG
Catalog No. 502706482
Encompass_Preferred
MW 382 Da, Purity >98%. ATP-sensitive potassium (KATP) channel blocker. Shows greater selectivity at KATP than at voltage-dependent and vascular inward rectifier K+ channels in smooth muscle cells. Active *in vivo* and *in vitro*.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6482 Supplier Abcam Supplier No. AB1416735MG
MW 382 Da, Purity >98%. ATP-sensitive potassium (KATP) channel blocker. Shows greater selectivity at KATP than at voltage-dependent and vascular inward rectifier K+ channels in smooth muscle cells. Active *in vivo* and *in vitro*.
The product is subject to the following: Abcam Restricted Use Statement