Learn More
Enzo Life Sciences U-46619 (10mg). CAS: 56985-40-1
SDP
Catalog No. 502011421
Encompass_Preferred
Prostaglandin H2/thromboxane A2 receptor agonist; stable analog of thromboxane A2. Induces platelet aggregation at 32nM and activates p38 MAPK. Alternative name: 9,11-Dideoxy-9alpha,11alpha-methanoepoxy prostaglandin F2alpha, 9,11-Dideoxy-9alpha,11alpha-methanoepoxy prosta5Z,13E-dien-1-oic acid. Purity: ≥98% (HPLC). Solubility: Soluble in 100% ethanol, DMSO, dimethyl formamide (100mg/ml) or PBS, pH 7.2 (2mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-1421 Supplier Enzo Life Sciences Supplier No. BMLPG0230010
Add to Cart
Add to Cart
Prostaglandin H2/thromboxane A2 receptor agonist; stable analog of thromboxane A2. Induces platelet aggregation at 32nM and activates p38 MAPK. Alternative name: 9,11-Dideoxy-9alpha,11alpha-methanoepoxy prostaglandin F2alpha, 9,11-Dideoxy-9alpha,11alpha-methanoepoxy prosta5Z,13E-dien-1-oic acid. Purity: ≥98% (HPLC). Solubility: Soluble in 100% ethanol, DMSO, dimethyl formamide (100mg/ml) or PBS, pH 7.2 (2mg/ml). Long Term Storage: -20°C.