Learn More
Cayman Chemical U-46619, 56985-40-1, 1 mg
SDP
Supplier:  Cayman Chemical 164501
Encompass_Preferred
TXA2 receptor agonist; causes platelet shape change (EC50 ~6 nM) and aggregation (EC50 ~97 nM)
Catalog No. NC9312824