Learn More
Abcam U-46619, Thrombo x ane A2 receptor agonist, 1MG
Supplier:  Abcam AB1445401MG
Encompass_Preferred
MW 350.5 Da, Purity >98%. Potent thromboxane A2 agonist (EC50 = 35 nM). Activates p38, ERK-1 and ERK-2. Induces platelet aggregation and shows vasoconstrictive effects *in vivo.*.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2103049