Learn More
Medchemexpress LLC U-50488 hydrochloride | 67197-96-0 | >98.0% | 405.79 | C19H27Cl3N2O | 10 MG
SDP
Catalog No. 5000285450
Encompass_Preferred
(±)-U-50488 hydrochloride is the racemic hydrochloride salt of U-50488, used as a research reagent to study kappa-opioid receptor (KOR) pharmacology and related signaling pathways. It is supplied in small solid quantities suitable for in vitro and early screening applications.
  • Selective kappa-opioid receptor agonist for receptor pharmacology studies.
  • High purity (>98.0%) suitable for biochemical and cell-based assays.
  • Available in small solid quantities (e.g., 10 mg) for research screening.
  • Molecular weight 405.79 and formula C19H27Cl3N2O; CAS 67197-96-0.
  • Offered as solid and in DMSO solution formats for convenient dosing.
Catalog No. 50-002-85450 Supplier Medchemexpress LLC Supplier No. HY1599710MG
(±)-U-50488 hydrochloride is the racemic hydrochloride salt of U-50488, used as a research reagent to study kappa-opioid receptor (KOR) pharmacology and related signaling pathways. It is supplied in small solid quantities suitable for in vitro and early screening applications.
  • Selective kappa-opioid receptor agonist for receptor pharmacology studies.
  • High purity (>98.0%) suitable for biochemical and cell-based assays.
  • Available in small solid quantities (e.g., 10 mg) for research screening.
  • Molecular weight 405.79 and formula C19H27Cl3N2O; CAS 67197-96-0.
  • Offered as solid and in DMSO solution formats for convenient dosing.