Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (±)-U-50488 hydrochloride | 67197-96-0 | 99.7% | 405.79 | C19H27Cl3N2O | 10 MG
SDP

Catalog No. 5000285475
Encompass_Preferred

(±)-U-50488 hydrochloride is the racemic trans hydrochloride salt of U-50488, a selective kappa-opioid receptor (KOR) agonist used in preclinical pharmacology and neuroscience research. The compound is supplied as a high-purity solid and is also available as a 10 mM solution in DMSO for laboratory assays.

  • Selective kappa-opioid receptor (KOR) agonist for preclinical assays.
  • High purity (99.7%) suitable for research applications.
  • Available as solid and as 10 mM solution in DMSO.
  • White to light yellow solid appearance.
  • Recommended storage: solid at 4°C; in solvent at -80°C (6 months) or -20°C (1 month).
  • Molecular weight 405.79; formula C19H27Cl3N2O.

Catalog No. 50-002-85475 Supplier Medchemexpress LLC Supplier No. HY15997B10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(±)-U-50488 hydrochloride is the racemic trans hydrochloride salt of U-50488, a selective kappa-opioid receptor (KOR) agonist used in preclinical pharmacology and neuroscience research. The compound is supplied as a high-purity solid and is also available as a 10 mM solution in DMSO for laboratory assays.

  • Selective kappa-opioid receptor (KOR) agonist for preclinical assays.
  • High purity (99.7%) suitable for research applications.
  • Available as solid and as 10 mM solution in DMSO.
  • White to light yellow solid appearance.
  • Recommended storage: solid at 4°C; in solvent at -80°C (6 months) or -20°C (1 month).
  • Molecular weight 405.79; formula C19H27Cl3N2O.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.