Learn More
Enzo Life Sciences U-69593 (25mg). CAS: 96744-75-1
SDP
Supplier:  Enzo Life Sciences BMLN7050025
Encompass_Preferred
Selective κ-opioid agonist. Thought to be selective for the κ-1 subtype (KD=3.8nM in human cerebral cortex membranes). Immunomodulatory activity. Purity: ≥99% (TLC). Solubility: Soluble in 100% ethanol, 0.1N hydrogen chloride, or DMSO. Insoluble in water or 0.1N sodium hydroxide. Long Term Storage: -20°C.
Catalog No. 50-201-1176