Learn More
American Radiolabeled Chemicals Inc U 69593 3H 25UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ART 179125UC
Encompass
U-69593-phenyl-3,4-3H 25uCi in ethanol, 1 mCi/ml, S.A. 30-60 Ci/mmol, M.W. 356.5, Shipped in Dry Ice
Catalog No. 50-901-120