Learn More
Medchemexpress LLC HY-13419 10mg , U-73122 CAS:112648-68-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1341910MG
Encompass_Preferred
HY-13419 10mg U-73122 CAS: 112648-68-7 U-73122 is a phospholipase C (PLC) and 5-LO (5-lipoxygenase) inhibitor with an IC50of 1-2.1 µM for PLC. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0060