Learn More
Apexbio Technology LLC U-73122 112648-68-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063911
Encompass_Preferred
U-73122 (CAS 112648-68-7) is a phospholipase C (PLC) inhibitor utilized in biomedical research By selectively targeting PLC an enzyme responsible for hydrolyzing phosphatidylinositol 4 5-bisphosphate into intracellular messengers diacylglycerol and inositol-triphosphate U-73122 attenuates downstream signaling events such as protein kinase C activation and intracellular calcium release In vitro U-73122 inhibits recombinant human PLC- 2 with an IC50 of approximately 6 M while minimally affecting PLC- 1 3 and 4 isoforms In animal models U-73122 demonstrates anti-inflammatory properties reducing carrageenan-induced rat paw edema and TPA-induced mouse ear edema
Catalog No. 50-000-63911 Supplier Apexbio Technology LLC Supplier No. B34225.1
U-73122 (CAS 112648-68-7) is a phospholipase C (PLC) inhibitor utilized in biomedical research By selectively targeting PLC an enzyme responsible for hydrolyzing phosphatidylinositol 4 5-bisphosphate into intracellular messengers diacylglycerol and inositol-triphosphate U-73122 attenuates downstream signaling events such as protein kinase C activation and intracellular calcium release In vitro U-73122 inhibits recombinant human PLC- 2 with an IC50 of approximately 6 M while minimally affecting PLC- 1 3 and 4 isoforms In animal models U-73122 demonstrates anti-inflammatory properties reducing carrageenan-induced rat paw edema and TPA-induced mouse ear edema