Learn More
Enzo Life Sciences U-73122, (5mg), CAS Number: 112648-68-7
SDP
Supplier:  Enzo Life Sciences BMLST3910005
Encompass_Preferred
Inhibitor of PLC activation. Alternative Name: 1(6((17β3methoxyestra-1 3 5(10)trien17yl)amino)hexyl)1Hpyrrole2 5dione. Formula: C29H40N2O3. MW: 464.7. CAS Number: 112648-68-7. Purity: ≥98% (HPLC). Appearance: white to off-white solid. Solubility: Soluble in methylene chloride or chloroform. Slightly soluble in DMSO (2mg/ml) or 100% ethanol (1mg/ml), sparingly soluble in aqueous solutions. Use/Stability: U73122 is only sparingly soluble in aqueous media. Compounds are delivered to cells either after dissolving in organic solvents or complexing with serum proteins. Dissolve a weighed amount of U-73122 in methylene chloride or chloroform. Dispense in aliquots sufficient for one experiment into suitable vessels. Evaporate solvent with nitrogen. Stock solutions in DMSO or ethanol are stable for 2 months at -20°C. Discard solutions turned to pink (indicates loss of inhibitor activity). Dried aliquots prepared from chloroform solutions are stable for up to 1 month at -20°C.
Catalog No. NC0356305