Learn More
Medchemexpress LLC U-73343 | 142878-12-4 | 99.4% | 466.66 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10863050MG
Encompass_Preferred
U-73343, which works as a protonophore, is an inactive analog of U-73122 and can be used as a negative control. It dose-dependently inhibits acid secretion irrespective of the stimulant. U-73122 is a phospholipase C (PLC) and 5-LO (5-lipoxygenase) inhibitor with an IC50 of 1-2.1 μM for PLC.
  • Functions as a protonophore
  • Can be used as a negative control
  • Inhibits acid secretion dose-dependently
Catalog No. 50-003-44888