Learn More
Enzo Life Sciences U-74389G (100mg). CAS: 153190-29-5
SDP
Catalog No. 502010826
Encompass_Preferred
Lazaroid inhibitor of iron-dependent lipid peroxidation. Antioxidant. Inhibits lipid peroxidation in a variety of experimental systems including cultured brain microvessel endothlial cells, Cu2+ treated monocyclic THP1 cells and LLC-PK1 cell layers where it blocks F2 isoprostane production. Prevented organ damage in intestinal cold storage, preservation and reperfusion injury. Alternative name: 21-(4-(2,6-di-1-Pyrrolidinyl-4-pyrimidinyl)-1-piperazinyl)-pregna-1,4,9(11)-triene-3,20-dione . (Z)-2-butenedioate. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (20mg/ml). Insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-0826 Supplier Enzo Life Sciences Supplier No. BMLFR1000100
Lazaroid inhibitor of iron-dependent lipid peroxidation. Antioxidant. Inhibits lipid peroxidation in a variety of experimental systems including cultured brain microvessel endothlial cells, Cu2+ treated monocyclic THP1 cells and LLC-PK1 cell layers where it blocks F2 isoprostane production. Prevented organ damage in intestinal cold storage, preservation and reperfusion injury. Alternative name: 21-(4-(2,6-di-1-Pyrrolidinyl-4-pyrimidinyl)-1-piperazinyl)-pregna-1,4,9(11)-triene-3,20-dione . (Z)-2-butenedioate. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (20mg/ml). Insoluble in water. Long Term Storage: -20°C.