Learn More
Abcam U-74389G, Antio x idant, 100MG
Catalog No. 502706497
Encompass_Preferred
MW 726.9 Da, Purity >98%. Antioxidant. Protects against lipid peroxidation by inhibiting the activity of the inducible nitric oxide synthase. Blood brain barrier permeable. Active both in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6497 Supplier Abcam Supplier No. AB141614100MG
MW 726.9 Da, Purity >98%. Antioxidant. Protects against lipid peroxidation by inhibiting the activity of the inducible nitric oxide synthase. Blood brain barrier permeable. Active both in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement