Learn More
Enzo Life Sciences U-75302 (100 µg). CAS: 119477-85-9
SDP
Catalog No. 502011704
Encompass_Preferred
Selective leukotriene B4 antagonist, Ki=159 nM for inhibition of [3H]-LTB4 binding to guinea pig lung membranes. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (25 mg/ml) or ethanol (25 mg/ml). Long Term Storage: -80°C.
Catalog No. 50-201-1704 Supplier Enzo Life Sciences Supplier No. BMLRA1020100
Selective leukotriene B4 antagonist, Ki=159 nM for inhibition of [3H]-LTB4 binding to guinea pig lung membranes. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (25 mg/ml) or ethanol (25 mg/ml). Long Term Storage: -80°C.