Learn More
Enzo Life Sciences U-83836E (100mg). CAS: 122003-11-6
SDP
Supplier:  Enzo Life Sciences BMLFR1010100
Encompass_Preferred
The lazaroid U-83836E prevents hydrogen peroxide-induced F2 isoprostane production in renal proximal tubular cells and reduces secondary brain injury in a rabbit model of cryogenic trauma. Alternative name: (-)-2-((4-(2,6-di-1-Pyrrolidinyl-4-pyrimidinyl)-1-piperazinyl)methyl)-3,4-dihydro-2,5,7,8-tetramethyl-2H-1-benzopyran-6-ol . 2HCl. Purity: ≥98% (HPLC). Solubility: Soluble in water (30 mg/ml) or ethanol (>50 mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0827