Learn More
Abcam U-83836E, Lazaroid antio x idant, 100MG
Catalog No. 502706496
Encompass_Preferred
MW 593.6 Da, Purity >98%. Lazaroid antioxidant, a steroid compound that inhibits lipid peroxidation. Inhibits cell proliferation (IC₅₀ = 6.30 mM). Potent free radical scavenger. Significantly protects against LPS induced lethality. Reduces secondary brain injury in a rabbit model of cryogenic trauma. Active in vivo and in vitro..
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6496 Supplier Abcam Supplier No. AB141615100MG
MW 593.6 Da, Purity >98%. Lazaroid antioxidant, a steroid compound that inhibits lipid peroxidation. Inhibits cell proliferation (IC₅₀ = 6.30 mM). Potent free radical scavenger. Significantly protects against LPS induced lethality. Reduces secondary brain injury in a rabbit model of cryogenic trauma. Active in vivo and in vitro..
The product is subject to the following: Abcam Restricted Use Statement