Learn More
Medchemexpress LLC U-99194 (maleate) | 234757-41-6 | 99.2% | 25 MG
SDP
Catalog No. 5000341852
Encompass_Preferred
U-99194 maleate is a selective, potent dopamine D3 receptor antagonist with a Kᵢ of 160 nM. It inhibits the activation of the D3 receptor mediated by endogenously released dopamine or exogenous D3 agonists. This compound can be used for the study of dopamine D3 receptor-mediated motor disorders, particularly kinetic tremors.
  • Selective, potent dopamine D3 receptor antagonist
  • Inhibits activation of D3 receptor
  • Abrogates the IPSC-suppressive effect of D3 agonists
  • Significantly suppresses Nicotine-induced tremor in mice
  • Useful for studying dopamine D3 receptor-mediated motor disorders
Catalog No. 50-003-41852 Supplier Medchemexpress LLC Supplier No. 25MGHY12701A
U-99194 maleate is a selective, potent dopamine D3 receptor antagonist with a Kᵢ of 160 nM. It inhibits the activation of the D3 receptor mediated by endogenously released dopamine or exogenous D3 agonists. This compound can be used for the study of dopamine D3 receptor-mediated motor disorders, particularly kinetic tremors.
  • Selective, potent dopamine D3 receptor antagonist
  • Inhibits activation of D3 receptor
  • Abrogates the IPSC-suppressive effect of D3 agonists
  • Significantly suppresses Nicotine-induced tremor in mice
  • Useful for studying dopamine D3 receptor-mediated motor disorders