Learn More
Medchemexpress LLC U-99194 maleate | 234757-41-6 | MFCD01529985 | 99.2% | 393.47 g/mol | C21H31NO6 | 5 MG
SDP
Catalog No. 5000263956
Encompass_Preferred
U-99194 maleate is a selective dopamine D3 receptor antagonist supplied as the maleate salt for research use. It is commonly used to probe D3-mediated signaling and motor disorder models, and is offered in small, research-sized quantities with reported high purity for biochemical and pharmacological studies.
  • Selective dopamine D3 receptor antagonist useful for receptor pharmacology studies.
  • Supplied as the maleate salt for stability and handling.
  • High reported purity (99.2%) suitable for biochemical assays.
  • Available in small milligram pack sizes for preclinical research.
  • Characterized by molecular formula C21H31NO6 and molecular weight 393.47 g/mol.
Catalog No. 50-002-63956 Supplier Medchemexpress LLC Supplier No. HY12701A5MG
U-99194 maleate is a selective dopamine D3 receptor antagonist supplied as the maleate salt for research use. It is commonly used to probe D3-mediated signaling and motor disorder models, and is offered in small, research-sized quantities with reported high purity for biochemical and pharmacological studies.
  • Selective dopamine D3 receptor antagonist useful for receptor pharmacology studies.
  • Supplied as the maleate salt for stability and handling.
  • High reported purity (99.2%) suitable for biochemical assays.
  • Available in small milligram pack sizes for preclinical research.
  • Characterized by molecular formula C21H31NO6 and molecular weight 393.47 g/mol.