Learn More
TARGETMOL CHEMICALS INC U 99194 maleate 2MG
SDP
Catalog No. 502876280
Encompass_Preferred
Also available in 1 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. U 99194 maleate (U-99194A maleate) is a D3 antagonist and increases social behaviors of isolation-induced aggressive male mice. Purity 99.45%
Catalog No. 50-287-6280 Supplier TARGETMOL CHEMICALS INC Supplier No. T234832MG
Also available in 1 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. U 99194 maleate (U-99194A maleate) is a D3 antagonist and increases social behaviors of isolation-induced aggressive male mice. Purity 99.45%